Learn More
ACROBiosystems Recombinant Protein;Cynomolgus / Rhesus macaque PD-L1 / B7-H1 Protein;HEK293;100UG;PD1,PDL1,PD-L1,CD274,B7-H1,B7H1,B7-H,B7H,PDCD1L1,PDCD1LG1
Supplier:  ACROBiosystems PD1C52H4100UG
Encompass_Preferred
Recombinant Protein;Cynomolgus / Rhesus macaque PD-L1 / B7-H1 Protein;100UG;Cynomolgus / Rhesus macaque PD-L1, His Tag (PD1-C52H4) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Arg 238 (Accession # G7PSE7).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.1 kDa. The protein migrates as 35-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus / Rhesus macaque PD-L1, His Tag (Cat. No. PD1-C52H4) at 1 μg/mL (100 μL/well) can bind MPDL3280A with a linear range of 0.4-3 ng/mL (QC tested).
Catalog No. 50-101-8403
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.