Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Human PLGF / PGF Protein;HEK293;20UG;PGF,PGF,PLGF,PLGF1,PLGF-1,PlGF2,PlGF,PGFL,PlGF-2,SHGC-10760  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner PGFH522920UG
Recombinant Protein;Human PLGF / PGF Protein;20UG;Human PLGF, His Tag (PGF-H5229) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Arg 170 (Accession # NP_002623.2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 20.2 kDa. The protein migrates as 28-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Rhesus macaque VEGF R1, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. VE1-R5257) at 5 μg/mL (100 μL/well) can bind Human PLGF, His Tag (Cat. No. PGF-H5229) with a linear range of 5-40 ng/mL (QC tested).;Immobilized Human PLGF, His Tag (Cat. No. PGF-H5229) at 1 μg/mL (100 μL/well) can bind Biotinylated Human VEGF R1, His Tag, Avi Tag (Cat. No. VE1-H82E3) with a linear range of 2-20 ng/mL (Routinely tested).
Catalog No. 50-101-8450