Learn More
ACROBiosystems Recombinant Protein;Human PH20 / SPAM1 Protein;HEK293;25UG;PH0,SPAM1,PH-20,SPAM-1,PH20,HYAL3,HYAL-3,HYA1,HYAL1,HYAL5,SPAG15
Supplier: ACROBiosystems PH0H522525UG
Recombinant Protein;Human PH20 / SPAM1 Protein;25UG;Human PH20, His Tag (PH0-H5225) is expressed from human 293 cells (HEK293). It contains AA Leu 36 - Tyr 482 (Accession # AAH26163).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 52.1 kDa. The protein migrates as 64-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.