Learn More
ACROBiosystems Recombinant Protein;Biotinylated Recombinant Protein L, ultra sensitivity (primary amine labeling, long spacer);E.coli;500UG;RPL,RPL,Protein L,ProteinL,IgG,IgM,IgE,IgA,IgD,antibody,Fc,RPL-P8141
Manufacturer:  ACROBiosystems RPLP814R500UG
Encompass_Preferred
Recombinant Protein;Biotinylated Recombinant Protein L, ultra sensitivity (primary amine labeling, long spacer);500UG;MABSol Biotinylated Recombinant Protein L, Superior Sensitivity (RPL-P814R) is expressed from E.coli cells. It is the biotinylated form of Recombinant Protein L (RPL-P3141). ;Immobilized Human IgG1 containing Kappa L chain at 5 g/mL (100 L/well) can bind Biotinylated Recombinant Protein L, Superior Sensitivity (Cat. No. RPL-P814R) with a linear range of 0.25-5 ng/mL (QC tested).;293 cells were transfected with anti-BCMA-scFv and RFP tag. 2e5 of the cells were first stained with Biotinylated Recombinant Protein L, Superior Sensitivity (Cat. No. RPL-P814R, 20 g/ml), followed by FITC Streptavidin. FITC Streptavidin was used as negative control.
Catalog No. 50-101-8513