Learn More
ACROBiosystems Recombinant Protein;Human SLAMF7 / CRACC / CD319 Protein, Fc Tag;HEK293;100UG;SL7,SLAMF7,CD319,CS1,CRACC,19A,FOAP-12,SLAMF7,FOAP12
Supplier:  ACROBiosystems SL7H5256100UG
Encompass_Preferred
Recombinant Protein;Human SLAMF7 / CRACC / CD319 Protein, Fc Tag;100UG;Human SLAMF7, Fc Tag (SL7-H5256) is expressed from human 293 cells (HEK293). It contains AA Ser 23 - Met 226 (Accession # AAH27867).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 49 kDa. The protein migrates as 60-67 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-101-8599