Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human CD47, Fc Tag, Avi Tag (Avitag™);HEK293;25UG;CD7,CD47,MER6,IAP,OA3
Supplier: ACROBiosystems CD7H82F625UG
Recombinant Protein;Biotinylated Human CD47, Fc Tag, Avi Tag (Avitag™);25UG;MABSol Biotinylated Human CD47, Fc Tag (CD7-H82F6) is expressed from human HEK293 cells. It contains AA Gln 19 - Pro 139 (Accession # NP_942088).;This protein carries a human IgG1 Fc fragment at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 42.2 kDa. The protein migrates as 50 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation.;Immobilized Human SIRP alpha, His Tag, Strep Tag (Cat. No. SIA-H5225) at 2 g/mL (100 L/well) can bind Biotinylated Human CD47, Fc Tag (Cat. No. CD7-H82F6) with a linear range of 0.008-0.25 g/mL (QC tested).;Immobilized Biotinylated Human CD47, Fc Tag (Cat. No. CD7-H82F6) at 1 g/mL (100 L/well) on streptavidin precoated (0.2ug/well) plate, can bind Cynomolgus SIRP alpha (Cat. No. SIA-C52H7) with a linear range of 0.039-0.625 g/mL (Routinely tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.