Learn More
ACROBiosystems Recombinant Protein;Human IgG1 Fc Protein, His Tag;HEK293;200UG;IG1,IgG1 Fc,IgG2,IgG3,IgG4,IgG2a,IgG Fc,IgG1
Supplier:  ACROBiosystems IG1H5225200UG
Encompass_Preferred
Recombinant Protein;Human IgG1 Fc Protein, His Tag;200UG;Human IgG Fc, His Tag (IG1-H5225) is expressed from human 293 cells (HEK293). It contains AA Glu 99 - Lys 330 (Accession # P01857).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27 kDa. The protein migrates as 34 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-108-9609
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.