Learn More
ACROBiosystems Recombinant Protein;Human Integrin alpha V beta 3 (ITGAV&ITGB3) Heterodimer Protein;HEK293;100UG;IT3,Integrin alpha V beta 3,ITGAV&ITGB3,Integrin,ITGAV,ITGA,ITGB3,ITGB,ITGAVB3
Supplier:  ACROBiosystems IT3H52E3100UG
Encompass_Preferred
Recombinant Protein;Human Integrin alpha V beta 3 (ITGAV&ITGB3) Heterodimer Protein;100UG;Human ITGAV&ITGB3, His Tag (IT3-H52E3) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) & Gly 27 - Asp 718 (ITGB3) (Accession # NP_002201 (ITGAV) & AAI27667.1 (ITGB3)).;Human ITGAV & ITGB3 Heterodimer Protein, produced by co-expression of ITGAV and ITGB3, has a calculated MW of 113 kDa (ITGAV) and 81.8 kDa (ITGB3). Subunit ITGAV is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB3 contains no tag but a basic tail at the C-terminus. The predicted N-terminus is Phe 31 (ITGAV) & Gly 27 (ITGB3). The reducing (R) protein migrates as 145 kDa (ITGAV) and 100-116 kDa (ITGB3) respectively due to glycosylation.;Immobilized Human ITGAVB3, His Tag (Cat. No. IT3-H52E3) at 5 μg/mL (100 μL/well) can bind Biotinylated TGF-Beta 1, N-His with a linear range of 0.2-5 μg/mL (QC tested).;Immobilized Human Fibronectin at 5μg/mL (100μL/we
Catalog No. 50-108-9643