Learn More
ACROBiosystems Recombinant Protein;Biotinylated Mouse TNF-alpha (HPLC-verified), Avi Tag (Avitag™);HEK293;25UG;TNA,TNF,DIF,TNF-alpha,TNFalpha,TNFA,TNFSF2,TNF-A,cachexin,cachectin,TNF-,TNF
Supplier:  ACROBiosystems TNAM82E925UG
Encompass_Preferred
Recombinant Protein;Biotinylated Mouse TNF-alpha (HPLC-verified), Avi Tag (Avitag™);25UG;MABSol Biotinylated Mouse TNF-alpha Protein, His Tag (TNA-M82E9) is expressed from human HEK293 cells. It contains AA Leu 80 - Leu 235 (Accession # P06804).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag (Avitag™) The protein has a calculated MW of 20.2 kDa. The protein migrates as 21 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation. More than 90% of Biotinylated Mouse TNF-alpha Protein, His Tag (TNA-M82E9) presents as active trimers with a molecular mass of 60 kDa, as determined by SEC-HPLC analysis.;Immobilized Human TNFR1, Fc Tag (Cat. No. TN1-H5251) at 5 g/mL (100 l/well) can bind Biotinylated Mouse TNF-alpha Protein, His Tag (Cat. No. TNA-M82E9) with a linear range of 0.8-12.5 ng/mL (QC tested).
Catalog No. 50-108-9747