Learn More
ACROBiosystems Recombinant Protein;Mouse PD-1 / PDCD1 Protein;HEK293;100UG;PD1,PDCD1,PD1,CD279,SLEB2,PDCD-1,PD-1,SLEB-2
Manufacturer: ACROBiosystems PD1M5228100UG
Recombinant Protein;Mouse PD-1 / PDCD1 Protein;100UG;Mouse PD-1, His Tag (PD1-M5228) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # NP_032824).;Mouse PDCD1 /PD1 is fused with a polyhistidine tag at the C-terminus, and has a calculated MW of 16.9 kDa. The N-terminus Sequence Analysis is Leu 25. The reducing (R) protein migrates as 25-45 kDa in SDS-PAGE due to different glycosylation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.