Learn More
ACROBiosystems Recombinant Protein;Human HVEM / TNFRSF14 Protein;HEK293;100UG;HVM,TNFRSF14,ATAR,HVEA,HVEM,LIGHTR,TR2,CD270
Supplier:  ACROBiosystems HVMH52E9100UG
Encompass_Preferred
Recombinant Protein;Human HVEM / TNFRSF14 Protein;100UG;Human HVEM, His Tag (HVM-H52E9) is expressed from human 293 cells (HEK293). It contains AA Leu 39 - Val 202 (Accession # NP_003811).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.2 kDa. The protein migrates as 33-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human LIGHT, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. LIT-H5256) at 2 μg/mL (100 μL/well) can bind Human HVEM, His Tag (Cat. No. HVM-H52E9) with a linear range of 0.04-0.6 μg/mL (QC tested).;Immobilized Human HVEM, His Tag (Cat.No. HVM-H52E9) at 5 μg/mL (100 μL/well) can bind Human BTLA (31-150), Fc Tag (Cat.No. BTA-H5256) with a linear range of 0.08-1.25 μg/mL (Routinely tested).
Catalog No. 50-113-8257