Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Human Nectin-3 / CD113 Protein, Fc Tag;HEK293;100UG;PV3,CD113,Nectin-3,PVRL3,Nectin
Manufacturer:  ACROBiosystemsSupplier Diversity Partner PV3H5255100UG
Encompass_Preferred
Recombinant Protein;Human Nectin-3 / CD113 Protein, Fc Tag;100UG;Human Nectin-3, Fc Tag (PV3-H5255) is expressed from human 293 cells (HEK293). It contains AA Gly 58 - Asp 400 (Accession # Q9NQS3).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 64.3 kDa. The protein migrates as 80-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Nectin-1, His Tag (Cat. No. PV1-H5223) at 2 μg/mL (100 μL/well) can bind Human Nectin-3, Fc Tag (Cat. No. PV3-H5255) with a linear range of 5-78 ng/mL (QC tested).;Immobilized Human CD113, Fc Tag (Cat. No. PV3-H5255) at 10 μg/mL (100 μL/well) can bind Biotinylated Human TIGIT, His Tag (Cat. No. TIT-H82E5) with a linear range of 0.3-5 μg/mL (Routinely tested).
Catalog No. 50-113-8272