Learn More
ACROBiosystems Recombinant Protein;Mouse 4-1BB / TNFRSF9 Protein;HEK293;100UG;41B,TNFRSF9,4-1BB,CD137,CDw137,ILA,TNFRSF-9,41BB,4-1BBL
Supplier:  ACROBiosystems 41BM52H7100UG
Encompass_Preferred
Recombinant Protein;Mouse 4-1BB / TNFRSF9 Protein;100UG;Mouse 4-1BB, His Tag (41B-M52H7) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Leu 211 (Accession # NP_001070976).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22 kDa. The protein migrates as 35-40 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Mouse 4-1BB, His Tag (Cat. No. 41B-M52H7) at 0.5μg/mL (100 µl/well),can bind Mouse 4-1BB Ligand , Fc Tag (Cat. No. 41L-M5257) with a linear of 0.1-3 ng/mL (QC tested).
Catalog No. 50-113-8283