Learn More
ACROBiosystems Recombinant Protein;Mouse IL-10 Protein;HEK293;20UG;IL0,CSIF,IL-10,IL10,IL10A,TGIF,B-TCGF,GVHDS,MGC126450,MGC126451,RP11-262N9.1,Interleukin 10,Interleukin-10,Interleukin10
Supplier:  ACROBiosystems IL0M424820UG
Encompass_Preferred
Recombinant Protein;Mouse IL-10 Protein;20UG;Mouse IL-10, His Tag (IL0-M4248) is expressed from human 293 cells (HEK293). It contains AA Ser 19 - Ser 178 (Accession # NP_034678).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 19.6 kDa. The protein migrates as 21 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-113-8377