Learn More
ACROBiosystems Recombinant Protein;Human EGFRvIII Protein;HEK293;50UG;EGI,EGFRvIII
Supplier:  ACROBiosystems EGIH52H450UG
Encompass_Preferred
Recombinant Protein;Human EGFRvIII Protein;50UG;Human EGFRvIII (Cat. No. EGI-H52H4) is expressed from human HEK293 cells. It contains AA Leu 25 - Ser 645 (30-297: Val Cys Gln...Cys Pro Arg → Gly) (Accession # NP_005219.2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 40.5 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cetuximab at 2 μg/mL (100 μL/well) can bind Human EGFRvIII, His Tag (Cat. No. EGI-H52H4) with a linear range of 0.2-5 ng/mL (QC tested).
Catalog No. 50-113-8401