Learn More
ACROBiosystems Recombinant Protein;Unconjugated Human CD47 Protein, His Tag & Avi Tag;HEK293;50UG;CD7,CD47,MER6,IAP,OA3
Supplier:  ACROBiosystems CD7HA2E950UG
Encompass_Preferred
Recombinant Protein;Unconjugated Human CD47 Protein, His Tag & Avi Tag;50UG;Human CD47, His Tag, Avi Tag (CD7-HA2E9) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 139 (Accession # NP_942088).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 16.9 kDa. The protein migrates as 33-44 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human SIRP alpha, Fc Tag (Cat. No. SIA-H5251) at 2μg/mL (100 μL/well) can bind Unconjugated Human CD47, His Tag, Avi Tag (Cat. No. CD7-HA2E9) with a linear range of 2-12.8 ng/mL (QC tested).;Immobilized Anti-CD47 MAb, Human IgG4 at 2μg/mL (100 μL/well) can bind Unconjugated Human CD47, His Tag, Avi Tag (Cat. No. CD7-HA2E9) with a linear range of 0.05-2.05 ng/mL (QC tested).
Catalog No. 50-125-3829