Learn More
Invitrogen™ iBind™ Flex Replacement Well Inserts Available on GSA/VA Contract for Federal Government customers only. Non-distribution product as customer accommodation.
For use with iBind™ Flex Western Blot Device
$159.00
Specifications
For Use With (Application) Western Blotting
View More Specs
Products 1
Catalog Number Mfr. No. Product Type Includes Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Includes Price Quantity & Availability  
SLF2002 Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only. View Documents Invitrogen™
SLF2002
Mini well insert with two wells on row 1 and 3 Two wells on row 1 and 3 Each for $159.00
Add to cart
Description
Description
The iBind™ Flex Western System offers automated western blot processing. Using customized well inserts, it can accommodate various blot formats including midi, mini, and vertically cut strips. Based on Sequential Lateral Flow (SLF) technology, all blocking, antibody incubation and washing steps during immunodetection are hands-free. Compared to manual methods, the iBind Flex Western System requires less primary antibody, enables reproducible results and is compatible with downstream detection protocols.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Western Blotting