Learn More
ACROBiosystems Recombinant Protein;Human / Cynomolgus / Rhesus macaque CD28 Protein;HEK293;100UG;CD8,CD28,Tp44
Manufacturer:  ACROBiosystems CD8H52HC100UG
Encompass_Preferred
Recombinant Protein;Human / Cynomolgus / Rhesus macaque CD28 Protein;100UG;Human / Cynomolgus / Rhesus macaque CD28 (CD8-H52Hc) is expressed from human 293 cells (HEK293). It contains AA Asn 19 - Pro 152 (Accession # P10747-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 17.0 kDa. The protein migrates as 30-42 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-CD28 MAb at 5 μg/mL (100 μL/well) can bind Human / Cynomolgus / Rhesus macaque CD28 (Cat. No. CD8-H52Hc) with a linear range of 0.3-2 ng/mL (QC tested).
Catalog No. 50-165-7505