Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Biotinylated Human BTLA (31-150) Protein, His Tag, Avi Tag (Avitag™);HEK293;25UG;BTA,BTLA,CD272
Manufacturer:  ACROBiosystemsSupplier Diversity Partner BTAH82E625UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human BTLA (31-150) Protein, His Tag, Avi Tag (Avitag™);25UG;Biotinylated Human BTLA (31-150), His Tag, Avi Tag (BTA-H82E6) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Ser 150 (Accession # AAP44003.1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 17.4 kDa. The protein migrates as 28-42 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human HVEM, mouse IgG2a Fc Tag, low endotoxin (Cat. No. HVM-H5255) at 5 μg/mL (100 μL/well) can bind Biotinylated Human BTLA (31-150), His Tag, Avi Tag (Cat. No. BTA-H82E6) with a linear range of 0.156-2.5 μg/mL (QC tested).
Catalog No. 50-165-7569