Learn More
ACROBiosystems Recombinant Protein;FITC-Labeled Human Siglec-2 / CD22 Protein;HEK293;25UG;SI2,CD22,SIGLEC2,BL-CAM,SIGLEC-2,Siglec2,Siglec-2,SIGLEC2FLJ22814,T-cell surface antigen Leu-14
Supplier: ACROBiosystems SI2HF2H625UG
Recombinant Protein;FITC-Labeled Human Siglec-2 / CD22 Protein;25UG;FITC-Labeled Human Siglec-2 (Cat. No. SI2-HF2H6) is expressed from human HEK293 cells. It contains AA Asp 20 - Arg 687 (Accession # P20273-1). It is the FITC labeled form of Human Siglec-2 (Cat. No. CD2-H52H8).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 77.0 kDa. The protein migrates as 100-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-Human CD22 MAb at 10 μg/mL (100 μL/well) can bind FITC-Labeled Human Siglec-2 (Cat. No. SI2-HF2H6) with a linear range of 0.625-10 μg/mL (Ex.488 nm/Em.535 nm) (QC tested).;293 cells were transfected with anti-CD22-scFv and RFP tag. 2e5 of the cells were stained with B. FITC-Labeled Human Siglec-2 / CD22 Protein (Cat. No. SI2-HF2H6, 10 µg/ml) and C. FITC-labeled Protein Control. A. Non-transfected 293 cells and C. FITC-labeled Protein Control were used as negative control. RFP was u
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.