Learn More
ACROBiosystems Recombinant Protein;Human FGF R4 / CD334 Protein;HEK293;200UG;FG4,FGFR4,FGFR-4,CD334,JTK2,MGC20292,TKF
Supplier:  ACROBiosystems FG4H5228200UG
Encompass_Preferred
Recombinant Protein;Human FGF R4 / CD334 Protein;200UG;Human FGF R4, His Tag (FG4-H5228) is expressed from human 293 cells (HEK293). It contains AA Leu 22 - Asp 369 (Accession # AAH11847).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 40.0 kDa. The protein migrates as 60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human FGF R4, His Tag (Cat. No. FG4-H5228) at 5 μg/mL (100 μL/well) can bind Biotinylated Human FGF basic, Tag Free (Cat. No. BFF-H8117) with a linear range of 20-156 ng/mL (QC tested).
Catalog No. 50-165-7591