Learn More
ACROBiosystems Recombinant Protein;Human IGF-II Protein, Fc Tag;HEK293;100UG;IG2,IGF2,IGF-2,C11orf43,FLJ22066,FLJ44734,IGF-II,IGFII,IGF-Ⅱ,IGFⅡ,PP9974
Supplier:  ACROBiosystems IG2H4260100UG
Encompass_Preferred
Recombinant Protein;Human IGF-II Protein, Fc Tag;100UG;Human IGF-II, Fc Tag (IG2-H4260) is expressed from human 293 cells (HEK293). It contains AA Ala 25 - Glu 91 (Accession # P01344-1).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 35 kDa. The protein migrates as 36 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Human IGFBP-3, His Tag (Cat. No. IG3-H5229) at 0.5 μg/mL (100 μL/well) can bind Human IGF-II, Fc Tag (Cat. No. IG2-H4260) with a linear range of 1-20 ng/mL (QC tested).
Catalog No. 50-165-7633