Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Human Neuroligin-4, Y-linked / NLGN4Y Protein;HEK293;50UG;NL1,NLGN4Y,neuroligin 4,Y-linked,NLGN4Y2
Manufacturer:  ACROBiosystemsSupplier Diversity Partner NL1H522850UG
Encompass_Preferred
Recombinant Protein;Human Neuroligin-4, Y-linked / NLGN4Y Protein;50UG;Human NLGN4Y, His Tag (NL1-H5228) is expressed from human 293 cells (HEK293). It contains AA Gln 44 - Ser 676 (Accession # Q8NFZ3-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 72.1 kDa. The protein migrates as 85-105 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7644