Learn More
ACROBiosystems Recombinant Protein;Cynomolgus IGF-I R / CD221 Protein;HEK293;100UG;IGR,IGF1R,CD221,IGFIR,IGFR,JTK13,IGF-IR,IGF-1R,JTK-13,MGC142170,MGC142172,MGC18216,IGF-I R
Supplier:  ACROBiosystems IGRC5225100UG
Encompass_Preferred
Recombinant Protein;Cynomolgus IGF-I R / CD221 Protein;100UG;Cynomolgus IGF-I R (IGR-C5225) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Asn 932 (Accession # G7P9I7-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 104.9 kDa, 81.1 kDa, 23.8 kDa. The protein migrates as 45-50 kDa, 96-115 kDa and 120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus IGF-I R (Cat. No. IGR-C5225) at 5 μg/mL (100 μL/well) can bind Human IGF-I, Fc Tag (Cat. No. IG1-H4269) with a linear range of 0.02-0.3 μg/mL (QC tested).
Catalog No. 50-165-7661