Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Rhesus macaque Siglec-2 / CD22 Protein;HEK293;100UG;SI2,CD22,SIGLEC2,BL-CAM,SIGLEC-2,Siglec2,Siglec-2,SIGLEC2FLJ22814,T-cell surface antigen Leu-14  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner SI2R52HA100UG
Recombinant Protein;Rhesus macaque Siglec-2 / CD22 Protein;100UG;Rhesus macaque Siglec-2 (SI2-R52Ha) is expressed from human 293 cells (HEK293). It contains AA Ser 20 - Arg 686 (Accession # EHH29920.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 76.9 kDa. The protein migrates as 95-120 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7735