Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human EpCAM / TROP1 Protein, Fc Tag, Avi Tag (Avitag™);HEK293;25UG;EPM,EPCAM,DIAR5,DIAR-5,EGP-2,EGP2,EGP314,EGP-314,EGP40,EGP-40,ESA,GA733-2,HNPCC8,HNPCC-8,KS1/4,KSA,M4S1,MIC18,MIC-18,MK-1,MK1,TACSTD1,TACSTD-1,TROP1,TROP-1,CD326,CD-326
Supplier: ACROBiosystems EPMH82F925UG
Recombinant Protein;Biotinylated Human EpCAM / TROP1 Protein, Fc Tag, Avi Tag (Avitag™);25UG;Biotinylated Human EpCAM, Fc Tag, Avi Tag (EPM-H82F9) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession # AAH14785.1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 56.2 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.