Learn More
ACROBiosystems Recombinant Protein;Human Transferrin Protein;HEK293;200UG;TRN,Transferrin,TF,DKFZp781D0156,PRO1557,PRO2086
Manufacturer:  ACROBiosystems TRNH4229200UG
Encompass_Preferred
Recombinant Protein;Human Transferrin Protein;200UG;Human Transferrin, His Tag (TRN-H4229) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Pro 698 (Accession # AAH59367).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 76 kDa. The protein migrates as 75-100 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Human Transferrin, His Tag (Cat. No. TRN-H4229) at 2 μg/mL (100 μL/well) can bind Biotinylated Human TfR, His Tag (Cat. No. TFR-H8243) with a linear range of 5-78 ng/mL (QC tested).
Catalog No. 50-165-7751