Learn More
ACROBiosystems Recombinant Protein;ActiveMax Recombinant Human TNF-alpha (HPLC-verified), low endotoxin;HEK293;25UG;TNA,TNF,DIF,TNF-alpha,TNFalpha,TNFA,TNFSF2,TNF-A,cachexin,cachectin,TNF-,TNF
Supplier:  ACROBiosystems TNAH421125UG
Encompass_Preferred
Recombinant Protein;ActiveMax Recombinant Human TNF-alpha (HPLC-verified), low endotoxin;25UG;Human TNF-alpha, Tag Free, low endotoxin (TNA-H4211) is expressed from human 293 cells (HEK293). It contains AA Val 77 - Leu 233 (Accession # P01375).;This protein carries no "tag". The protein has a calculated MW of 17.4 kDa. The protein migrates as 17 kDa and 18 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Human TNF-alpha, Tag Free (HPLC-verified), low endotoxin (Cat. No. TNA-H4211) at 2 g/mL (100 L/well) can bind Humira with a linear range of 0.2-2 ng/mL (QC tested).;Immobilized Human TNF-alpha, Tag Free (HPLC-verified), low endotoxin (Cat. No. TNA-H4211) at 2 g/mL (100 L/well) can bind Human TNFR1, Fc Tag (Cat. No. TN1-H5251) with a linear range of 0.2-3 ng/mL (Routinely tested).;Humira (Adalimumab) captured on CM5 chip via anti-human IgG Fc antibodies surface, can bind Human TNF-alpha, Tag Free (HPLC-verified), low endotoxin (Cat. No. TNA-H4211) with an aff
Catalog No. 50-165-7763
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.