Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Human MMP-1 Protein;HEK293;50UG;MM1,MMP1,MMP-1,CLG,CLGN,Matrix Metallopeptidase 1  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner MM1H522250UG
Recombinant Protein;Human MMP-1 Protein;50UG;Human MMP-1, His Tag (MM1-H5222) is expressed from human 293 cells (HEK293). It contains AA Phe 20 - Asn 469 (Accession # NP_002412.1).;rh MMP1 is fused with a polyhistidine tag at the C-terminus, and has a calculated MW of 52.7 kDa. The predicted N-terminus is Phe 20. The reducing (R) protein migrates as 52-55 kDa in SDS-PAGE .
Catalog No. 50-165-7805