Learn More
ACROBiosystems Recombinant Protein;Human CD9 Protein;HEK293;20UG;CD9,CD9,MIC3,TSPAN29,GIG2,MRP1,BTCC1,DRAP27,5H9,BTCC-1,DRAP-27,MIC-3,MRP-1,TSPAN-29,Tetraspanin-29,Tetraspanin 29
Supplier:  ACROBiosystems CD9H522020UG
Encompass_Preferred
Recombinant Protein;Human CD9 Protein;20UG;Human CD9, His Tag (CD9-H5220) is expressed from human 293 cells (HEK293). It contains AA Ser 112 - Ile 195 (Accession # P21926-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 11.6 kDa. The protein migrates as 12 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-7866