Learn More
ACROBiosystems Recombinant Protein;Human CD38 Protein, Llama IgG2b Fc Tag, low endotoxin;HEK293;50UG;CD8,CD38,T10,cADPr hydrolase 1
Supplier:  ACROBiosystems CD8H525250UG
Encompass_Preferred
Recombinant Protein;Human CD38 Protein, Llama IgG2b Fc Tag, low endotoxin;50UG;Human CD38, Llama IgG2b Fc Tag, low endotoxin (CD8-H5252) is expressed from human 293 cells (HEK293). It contains AA Val 43 - Ile 300 (Accession # P28907-1).;This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 57.7 kDa. The protein migrates as 66-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD38, Llama IgG2b Fc Tag, low endotoxin (Cat. No. CD8-H5252) at 1 μg/mL (100 μL/well) can bind Anti-CD38 MAb, Human IgG1 with a linear range of 0.4-6 ng/mL (QC tested).
Catalog No. 50-165-7909