Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Human LILRB4 / CD85k / ILT3 Protein, Mouse IgG2a Fc Tag, low endotoxin;HEK293;100UG;CDK,LILRB4,ILT3,LIR5,CD85K,HM18,LILRB-4,ILT-3,LIR-5
Manufacturer:  ACROBiosystemsSupplier Diversity Partner CDKH5259100UG
Encompass_Preferred
Recombinant Protein;Human LILRB4 / CD85k / ILT3 Protein, Mouse IgG2a Fc Tag, low endotoxin;100UG;Human LILRB4, Mouse IgG2a Fc Tag, low endotoxin (CDK-H5259) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Glu 259 (Accession # AAH26309.1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 53.2 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ANGPTL7, His Tag (Cat. No. AN7-H52H5) at 5 μg/mL (100 μL/well) can bind Human LILRB4, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CDK-H5259) with a linear range of 0.313-2.5 μg/mL (QC tested).
Catalog No. 50-165-7929