Learn More
ACROBiosystems Recombinant Protein;Marmoset OX40 / TNFRSF4 / CD134 Protein;HEK293;100UG;OX0,TNFRSF4,OX40,CD134,OX40L receptor,ACT35,TXGP1L,OX-40
Supplier:  ACROBiosystems OX0M522B100UG
Encompass_Preferred
Recombinant Protein;Marmoset OX40 / TNFRSF4 / CD134 Protein;100UG;Marmoset OX40 (OX0-M522b) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Ala 214 (Accession # F7DHA3-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.8 kDa. The protein migrates as 42-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human OX40 Ligand, Fc Tag (Cat. No. OXL-H526x) at 5 μg/mL (100 μL/well) can bind Marmoset OX40 (Cat. No. OX0-M522b) with a linear range of 0.078-1.25 μg/mL (QC tested).;Immobilized Marmoset OX40 (Cat. No. OX0-M522b) at 2 μg/mL (100 μL/well) can bind Rhesus macaque OX40 Ligand, Mouse IgG2a Fc Tag (Cat. No. OXL-R5259) with a linear range of 0.2-3 ng/mL (Routinely tested).
Catalog No. 50-165-8074