Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Recombinant Influenza A [A/Shanghai/2/2013(H7N9)] Hemagglutinin (HA);HEK293;100UG;HA9,HA,Hemagglutinin  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner HA9V5227100UG
Recombinant Protein;Recombinant Influenza A [A/Shanghai/2/2013(H7N9)] Hemagglutinin (HA);100UG;Influenza , His Tag (HA9-V5227) is expressed from human 293 cells (HEK293).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 58.6 kDa. The protein migrates as 63-75 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8105