Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Biotinylated Human CD4, His Tag & Avi Tag (Avitag™);HEK293;25UG;CD4,CD4,CD-4,CD4mut,CD-4mut,LEU3,LEU-3
Manufacturer:  ACROBiosystemsSupplier Diversity Partner CD4H82E825UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human CD4, His Tag & Avi Tag (Avitag™);25UG;Biotinylated Human CD4, His Tag, Avi Tag (CD4-H82E8) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Pro 396 (Accession # P01730-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 45.0 kDa. The protein migrates as 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized HIV GP120 (HIV), His Tag (Cat. No. GP5-V15224) at 5 μg/mL (100 μL/well) can bind Biotinylated Human CD4, His Tag, Avi Tag (Cat. No. CD4-H82E8) with a linear range of 2-39 ng/mL (QC tested).;Immobilized HIV GP120 (HIV), His Tag (Cat. No. GP4-V15223) at 5 μg/mL (100 μL/well) can bind Biotinylated Human CD4, His Tag, Avi Tag (Cat. No. CD4-H82E8) with a linear range of 2-78 ng/mL (Routinely tested).
Catalog No. 50-165-8190