Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Biotinylated Human CBLB / RNF56 Protein, His Tag & Avi Tag (Avitag™);E.coli;25UG;CBB,CBL-B,CBLB,RNF56  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner CBBH81Q825UG
Recombinant Protein;Biotinylated Human CBLB / RNF56 Protein, His Tag & Avi Tag (Avitag™);25UG;Biotinylated Human CBLB, His Tag (CBB-H81Q8) is expressed from E.coli cells. It contains AA Ala 39 - Phe 426 (Accession # Q13191-1).;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 47.8 kDa. The protein migrates as 45 kDa and 46 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8201