Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Human FABP6 Protein;E.coli;100UG;FA6,FABP6,ILBP,ILLBP,Gastrotropin,GT,I-15P,I-BABP,I-BALB,I-BAP  DFS Item
Manufacturer:  ACROBiosystemsSupplier Diversity Partner FA6H5141100UG
Recombinant Protein;Human FABP6 Protein;100UG;Human FABP6, His Tag (FA6-H5141) is expressed from E.coli cells. It contains AA Ala 2 - Ala 128 (Accession # AAH22489).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 15.3 kDa. The protein migrates as 15 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8248