Learn More
ACROBiosystems Recombinant Protein;Cynomolgus / Rhesus macaque CD47 Protein, His Tag;HEK293;50UG;CD7,CD47,MER6,IAP,OA3
Supplier: ACROBiosystems CD7C52H150UG
Recombinant Protein;Cynomolgus / Rhesus macaque CD47 Protein, His Tag;50UG;Cynomolgus / Rhesus macaque CD47, His Tag (CD7-C52H1) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Glu 141 (Accession # NP_001253446.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.8 kDa. The protein migrates as 30-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human SIRP alpha, Fc Tag (Cat. No. SIA-H5251) at 5 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque CD47, His Tag (Cat. No. CD7-C52H1) with a linear range of 4-63 ng/mL (QC tested).;Immobilized Anti-CD47, Human IgG4 at 2 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque CD47, His Tag (Cat. No. CD7-C52H1) with a linear range of 0.2-5 ng/mL (Routinely tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.