Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human M-CSF R / CSF1R / CD115 Protein, His Tag & Avi Tag (Avitag™);HEK293;25UG;CSR,CSF1R,C-FMS,CD115,CSFR,FIM2,FMS,M-CSFR,CSF-1R,CSF1 R
Supplier:  ACROBiosystems CSRH82E025UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human M-CSF R / CSF1R / CD115 Protein, His Tag & Avi Tag (Avitag™);25UG;Biotinylated Human M-CSF R, His Tag (CSR-H82E0) is expressed from human 293 cells (HEK293). It contains AA Ile 20 - Glu 512 (Accession # P07333-1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 57.1 kDa. The protein migrates as 65-92 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human M-CSF, His Tag (Cat. No. MCF-H5247) at 5 μg/mL (100 μL/well) can bind Biotinylated Human M-CSF R, His Tag (Cat. No. CSR-H82E0) with a linear range of 16-500 ng/mL (QC tested).
Catalog No. 50-165-8322