Learn More
ACROBiosystems Recombinant Protein;Human TNFSF11 / RANKL / CD254 Protein, His tag;HEK293;50UG;RAL,TNFSF11,RANKL,CD254,TRANCE,OPGL,ODF
Supplier:  ACROBiosystems RALH524050UG
Encompass_Preferred
Recombinant Protein;Human TNFSF11 / RANKL / CD254 Protein, His tag;50UG;Human TNFSF11, His Tag (RAL-H5240) is expressed from human 293 cells (HEK293). It contains AA Gly 64 - Asp 245 (Accession # AAC51762.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 23.1 kDa. The protein migrates as 30-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human RANK, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. RAK-H5251) at 0.5 μg/mL (100 μL/well) can bind Human TNFSF11, His Tag (Cat. No. RAL-H5240) with a linear range of 0.4-6 ng/mL (QC tested).
Catalog No. 50-165-8339