Learn More
ACROBiosystems Recombinant Protein;Cynomolgus / Rhesus macaque BCMA / TNFRSF17 Protein, Fc Tag;HEK293;100UG;BCA,TNFRSF17,CD269,BCM,BCMA,TNFRSF-17
Supplier:  ACROBiosystems BCAC5253100UG
Encompass_Preferred
Recombinant Protein;Cynomolgus / Rhesus macaque BCMA / TNFRSF17 Protein, Fc Tag;100UG;Cynomolgus / Rhesus macaque BCMA, Fc Tag (BCA-C5253) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 53 (Accession # G7Q0I4-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 32.1 kDa. As a result of glycosylation, the protein migrates as 37 kDa and 38-42 kDa under reducing (R) condition, and 60-66 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human BAFF, His Tag (Cat. No. BAF-H5248) at 5 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque BCMA, Fc Tag (Cat. No. BCA-C5253) with a linear range of 0.05-2 ng/mL (QC tested).
Catalog No. 50-165-8358