Learn More
ACROBiosystems Recombinant Protein;Human Osteoactivin / GPNMB Protein;HEK293;100UG;GPB,GPNMB,HGFIN,NMB,Osteoactivin,DC-HIL,OA
Supplier: ACROBiosystems GPBH5229100UG
Recombinant Protein;Human Osteoactivin / GPNMB Protein;100UG;Human GPNMB, His Tag (GPB-H5229) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Pro 486 (Accession # AAH32783).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 52.9 kDa. The protein migrates as 85-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.