Learn More
ACROBiosystems Recombinant Protein;Mouse LILRB4 / CD85k / ILT3 Protein, Fc Tag;HEK293;100UG;CDK,LILRB4,ILT3,LIR5,CD85K,HM18,LILRB-4,ILT-3,LIR-5
Supplier:  ACROBiosystems CDKM5250100UG
Encompass_Preferred
Recombinant Protein;Mouse LILRB4 / CD85k / ILT3 Protein, Fc Tag;100UG;Mouse LILRB4, Fc Tag (CDK-M5250) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Lys 238 (Accession # Q64281-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50.7 kDa. As a result of glycosylation, the protein migrates as 60-66 kDa under reducing (R) condition, and 120-140 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human ANGPTL7, His Tag (Cat. No. AN7-H52H5) at 5 μg/mL (100 μL/well) can bind Mouse LILRB4, Fc Tag (Cat. No. CDK-M5250) with a linear range of 0.156-1.25 μg/mL (QC tested).
Catalog No. 50-165-8391