Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Human Galectin-3 Protein;HEK293;100UG;GA3,Galectin-3,LGALS3,MAC2,Gal-3,35 kDa lectin,CBP 35,GALBP,IgE-binding protein,L-31,Laminin-binding protein,Lectin L-29,Mac-2 antigen,Galectin 3,LGALS-3,MAC-2,Gal3,L31,Galectin3
Manufacturer:  ACROBiosystemsSupplier Diversity Partner GA3H5247100UG
Encompass_Preferred
Recombinant Protein;Human Galectin-3 Protein;100UG;Human Galectin-3, His Tag (GA3-H5247) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ile 250 (Accession # P17931-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 27.5 kDa. The protein migrates as 35-41 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8407