Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human VSIG8 Protein, Fc Tag, Avi Tag (Avitag™);HEK293;25UG;VS8,VSIG8,C1orf204
Supplier:  ACROBiosystems VS8H82F225UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human VSIG8 Protein, Fc Tag, Avi Tag (Avitag™);25UG;Biotinylated Human VSIG8, Fc Tag (VS8-H82F2) is expressed from human 293 cells (HEK293). It contains AA Val 22 - Gly 263 (Accession # NP_001013683.1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 55.5 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8422