Learn More
ACROBiosystems Recombinant Protein;Rhesus macaque B7-H5 / Gi24 / VISTA Protein;HEK293;100UG;B75,B7-H5,C10orf54,Sisp-1,SISP1,Gi24,VISTA,B7H5
Supplier: ACROBiosystems B75C5220100UG
Recombinant Protein;Rhesus macaque B7-H5 / Gi24 / VISTA Protein;100UG;Rhesus macaque B7-H5, His Tag (B75-C5220) is expressed from human 293 cells (HEK293). It contains AA Phe 33 - Thr 192 (Accession # NP_001177807.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.9 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.