Learn More
ACROBiosystems Recombinant Protein;Human CD28H / IGPR-1 Protein, Fc Tag;HEK293;100UG;CDH,TMIGD2,IGPR-1,CD28H,IGPR1
Manufacturer:  ACROBiosystems CDHH5251100UG
Encompass_Preferred
Recombinant Protein;Human CD28H / IGPR-1 Protein, Fc Tag;100UG;Human CD28H, Fc Tag (CDH-H5251) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Gly 150 (Accession # AAH15655.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.6 kDa. As a result of glycosylation, the protein migrates as 45-60 kDa under reducing (R) condition, and 90-120 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-165-8451