Learn More
ACROBiosystems Recombinant Protein;Human VEGF R3 / FLT4 Protein, Fc Tag;HEK293;100UG;FL4,Flt-4,FLT4,LMPH1A,PCLFLT41,VEGFR3,VEGFR-3,FLT-4,FLT-41,FLT41,PCL,VEGF R3
Supplier: ACROBiosystems FL4H5251100UG
Recombinant Protein;Human VEGF R3 / FLT4 Protein, Fc Tag;100UG;Human VEGF R3, Fc Tag (FL4-H5251) is expressed from human 293 cells (HEK293). It contains AA Tyr 25 - Ile 776 (Accession # NP_002011.2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 111.2 kDa. As a result of glycosylation, the protein migrates as 140-160 kDa under reducing (R) condition, and 280-320 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human VEGF-C, His Tag (Cat. No. VEC-H4225) at 5μg/mL (100 μL/well) can bind Human VEGF R3, Fc Tag (Cat. No. FL4-H5251) with a linear range of 0.6-10 ng/mL (QC tested).;Immobilized Human VEGF R3, Fc Tag (Cat. No. FL4-H5251) at 5μg/mL (100 μL/well) can bind Human VEGF-D, His Tag (Cat. No. VED-H5228) with a linear range of 0.08-0.6 μg/mL (Routinely tested).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.