Learn More
ACROBiosystemsSupplier Diversity Partner Recombinant Protein;Human SCARB2 / SR-B2 / LIMPII Protein, Fc Tag;HEK293;100UG;SC2,SCARB2,CD36L2,LIMP2,LIMPII,LGP85,CD36,AMRF,EPM4,HLGP85,SR-BII,SCARB-2,CD36-L2,LIMP-2,
Manufacturer:  ACROBiosystemsSupplier Diversity Partner SC2H5250100UG
Encompass_Preferred
Recombinant Protein;Human SCARB2 / SR-B2 / LIMPII Protein, Fc Tag;100UG;Human SCARB2, Fc Tag (SC2-H5250) is expressed from human 293 cells (HEK293). It contains AA Arg 27 - Thr 432 (Accession # AAH21892).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 72.5 kDa. The protein migrates as 90-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8497