Learn More
ACROBiosystems Recombinant Protein;Human NKp46 / NCR1 / CD335 Protein, Fc Tag;HEK293;50UG;NC1,NCR1,LY94,CD335,NK-p46,NKp46,hNKp46,NCR-1,LY-94
Supplier:  ACROBiosystems NC1H525750UG
Encompass_Preferred
Recombinant Protein;Human NKp46 / NCR1 / CD335 Protein, Fc Tag;50UG;Human NKp46, Fc Tag (NC1-H5257) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asn 254 (Accession # AAH64806).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 53 kDa. The protein migrates as 53-75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8514